siRNA Details
Gene Symbol: 4933424G05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045372.1 | 5 | 878 | |||
GenBank mRNA | AK016892.1 | - | 5 | 879 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933424G05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045372.1 | 5 | 878 | |||
GenBank mRNA | AK016892.1 | - | 5 | 879 |