siRNA Details
Gene Symbol: 4933424G05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045372.1 | 2, 3 | 199 | |||
GenBank mRNA | AK016892.1 | - | 2, 3 | 200 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716571.1 | - | 2, 3 | 200 | Chr. 18 - 14988995 - 15047072 [+] on Build GRCm38 |
