siRNA Details
Gene Symbol: 1700093E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007049.1 | - | 1 | 603 | ||
AK018923.1 | - | 1 | 603 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707119.1 | - | 1 | 600 | Chr. 2 - 178417473 - 178418240 [-] on Build GRCm38 |
