siRNA Details
Gene Symbol: 4930552N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040661.1 | 2 | 163 | |||
GenBank mRNA | AK016098.1 | - | 2 | 164 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715700.1 | - | 2 | 164 | Chr. 4 - 52814604 - 52827972 [+] on Build GRCm38 |
