siRNA Details
Gene Symbol: 4930552F14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016097.1 | - | 2 | 424 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715699.1 | - | 2 | 424 | Chr. 16 - 43979343 - 43983351 [+] on Build GRCm38 |
