siRNA Details
Gene Symbol: Gm32548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042350.1 | - | 3 | 290 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB639252.1 | - | 3 | 290 | Chr. 18 - 73711326 - 73717202 [+] on Build GRCm38 |
