siRNA Details
Gene Symbol: 1700027H10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040594.1 | 4 | 457 | |||
GenBank mRNA | AK006417.1 | - | 4 | 458 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706472.1 | - | 4 | 455 | Chr. 3 - 45416584 - 45439307 [-] on Build GRCm38 |
