siRNA Details
Gene Symbol: 5031425E22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017143.1 | - | 382 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716854.1 | - | 3 | 382 | Chr. 5 - 23384984 - 23433311 [-] on Build GRCm38 |
