siRNA Details
Gene Symbol: 9030625N01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018580.1 | - | 1 | 797 | ||
AK035942.1 | - | 1 | 1364 | |||
AK076245.1 | - | 1 | 1367 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX210678.1 | - | 1 | 512 | Chr. 19 - 4626247 - 4627075 [+] on Build GRCm38 |
