siRNA Details
Gene Symbol: 8030423F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045738.1 | 3 | 333 | |||
GenBank mRNA | AK033101.1 | - | 3 | 334 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB624154.1 | - | 3 | 334 | Chr. 5 - 52609735 - 52619011 [-] on Build GRCm38 |
