siRNA Details
Gene Symbol: Gm17115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085908.1 | - | 1 | 642 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB518041.2 | - | 1 | 394 | Chr. 17 - 32788654 - 32789591 [+] on Build GRCm38 |
