siRNA Details
Gene Symbol: Gm16252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076831.1 | - | 4 | 587 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV264873.2 | - | 3 | 379 | Chr. 9 - 103472871 - 103492928 [+] on Build GRCm38 |
