siRNA Details
Gene Symbol: Gm19395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076832.1 | - | 3 | 591 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV264908.2 | - | 3 | 601 | Chr. 10 - 53280287 - 53283686 [+] on Build GRCm38 |
