siRNA Details
Gene Symbol: Gm5089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047660.1 | - | 2 | 188 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB647616.1 | - | 2 | 188 | Chr. 14 - 122365877 - 122406743 [-] on Build GRCm38 |
