siRNA Details
Gene Symbol: 4930532G15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089488.1 | - | 1 | 1084 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY764530.1 | - | 1 | 342 | Chr. 1 - 185361411 - 185362118 [-] on Build GRCm38 |
