siRNA Details
Gene Symbol: Gm36745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139415.1 | - | 2, 3 | 1285 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB271466.2 | - | 1, 2 | 390 | Chr. 9 - 113636532 - 113639889 [-] on Build GRCm38 |
