siRNA Details
Gene Symbol: 4930426I24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015206.1 | - | 1 | 421 | ||
AK020166.1 | - | 1 | 749 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF198157.1 | - | 1 | 344 | Chr. 12 - 76402738 - 76403578 [-] on Build GRCm38 |
