siRNA Details
Gene Symbol: 2610304O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011983.1 | - | 1 | 378 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711177.1 | - | 1 | 378 | Chr. 13 - 28725904 - 28726348 [-] on Build GRCm38 |
