siRNA Details
Gene Symbol: 4930597A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016402.1 | - | 2 | 58 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716054.1 | - | 2 | 58 | Chr. 11 - 44586317 - 44591229 [+] on Build GRCm38 |
