siRNA Details
Gene Symbol: 5330416C01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045624.1 | - | 3 | 357 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB644122.1 | - | 3 | 357 | Chr. 13 - 97743914 - 97747411 [-] on Build GRCm38 |
