siRNA Details
Gene Symbol: 2810019C22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012763.1 | - | 1 | 420 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711879.1 | - | 1 | 418 | Chr. 9 - 88448043 - 88448947 [-] on Build GRCm38 |
