siRNA Details
Gene Symbol: Gm16325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045949.1 | 1, 2 | 344 | |||
GenBank mRNA | AK139061.1 | - | 1, 2 | 345 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB639751.1 | - | 1, 2 | 345 | Chr. 3 - 146641932 - 146648300 [+] on Build GRCm38 |
