siRNA Details
Gene Symbol: 2900024D18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013581.1 | - | 1 | 494 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712542.1 | - | 1 | 494 | Chr. 13 - 6925283 - 6925851 [-] on Build GRCm38 |
