siRNA Details
Gene Symbol: Gm31359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052782.1 | - | 528 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB602732.2 | - | 3 | 528 | Chr. 3 - 85397523 - 85428128 [-] on Build GRCm38 |
