siRNA Details
Gene Symbol: 1700017I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006067.1 | - | 2 | 304 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706181.1 | - | 2 | 302 | Chr. 9 - 89802870 - 89806055 [+] on Build GRCm38 |
