siRNA Details
Gene Symbol: Fendrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087264.1 | - | 5 | 1804 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB534926.2 | - | 1 | 84 | Chr. 8 - 121054882 - 121059974 [-] on Build GRCm38 |
