siRNA Details
Gene Symbol: Gm38973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146287.1 | - | 2 | 426 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY738096.1 | - | 2 | 426 | Chr. 7 - 27649633 - 27651059 [-] on Build GRCm38 |
