siRNA Details
Gene Symbol: AW555355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK157022.1 | - | 1 | 2298 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AW825539.1 | - | 1 | 382 | Chr. 2 - 90569538 - 90570071 [-] on Build GRCm38 |
