siRNA Details
Gene Symbol: 2310007H11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009208.1 | - | 1 | 462 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709017.1 | - | 1 | 464 | Chr. 2 - 164522215 - 164523044 [+] on Build GRCm38 |
