siRNA Details
Gene Symbol: 1700012D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031468.1 | - | 1 | 937 | ||
AK047116.1 | - | 1 | 937 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV321778.2 | - | 1 | 262 | Chr. 7 - 111121322 - 111121995 [-] on Build GRCm38 |
