siRNA Details
Gene Symbol: 4930473H19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015566.1 | - | 3 | 462 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715116.1 | - | 3 | 461 | Chr. 12 - 88434190 - 88442219 [-] on Build GRCm38 |
