siRNA Details
Gene Symbol: Gm26761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033648.1 | - | 2 | 135 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB625243.1 | - | 2 | 135 | Chr. 5 - 64696987 - 64698520 [-] on Build GRCm38 |
