siRNA Details
Gene Symbol: 4732419C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076318.1 | - | 3 | 2260 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4732419C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076318.1 | - | 3 | 2260 |