siRNA Details
Gene Symbol: Gm31219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136039.1 | - | 3 | 633 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719968.1 | - | 3 | 630 | Chr. 13 - 76255949 - 76270964 [+] on Build GRCm38 |
