siRNA Details
Gene Symbol: 1700109G15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046197.1 | 1, 2 | 56 | |||
GenBank mRNA | AK018946.1 | - | 1, 2 | 57 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707234.1 | - | 1, 2 | 57 | Chr. 11 - 84716975 - 84720432 [-] on Build GRCm38 |
