siRNA Details
Gene Symbol: Gm36394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043176.1 | - | 2 | 2647 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB254921.2 | - | 1 | 610 | Chr. 19 - 8612226 - 8612899 [-] on Build GRCm38 |
