siRNA Details
Gene Symbol: 4933400C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040770.1 | 5 | 940 | |||
GenBank mRNA | AK077069.1 | - | 5 | 941 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933400C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040770.1 | 5 | 940 | |||
GenBank mRNA | AK077069.1 | - | 5 | 941 |