siRNA Details
Gene Symbol: Gm26644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144194.1 | - | 2 | 771 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY755223.1 | - | 1 | 479 | Chr. 19 - 44692357 - 44693035 [-] on Build GRCm38 |
