siRNA Details
Gene Symbol: A230077H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038939.1 | - | 655 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ443807.1 | - | 1 | 76 | Chr. 7 - 40851266 - 40875324 [-] on Build GRCm38 |
