siRNA Details
Gene Symbol: 4930544I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132687.1 | - | 383 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614671.1 | - | 3 | 383 | Chr. 12 - 90753840 - 90969768 [-] on Build GRCm38 |
