siRNA Details
Gene Symbol: Gm15834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053027.1 | - | 2 | 1683 | ||
AK138448.1 | - | 2 | 1687 | |||
AK144771.1 | - | 3 | 190 | |||
BB183984.2 | - | 1 | 217 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY734356.1 | - | 3 | 190 | Chr. 1 - 58505150 - 58516120 [+] on Build GRCm38 |
