siRNA Details
Gene Symbol: 2810427C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013171.1 | - | 1 | 788 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712193.1 | - | 1 | 794 | Chr. 2 - 92306175 - 92307120 [+] on Build GRCm38 |
