siRNA Details
Gene Symbol: 1700024F20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006298.1 | - | 1 | 45 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706376.1 | - | 1 | 39 | Chr. 9 - 118924824 - 118925376 [-] on Build GRCm38 |
