siRNA Details
Gene Symbol: 1700024B18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045479.1 | 4 | 342 | |||
GenBank mRNA | AK006293.1 | - | 4 | 343 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706372.2 | - | 4 | 342 | Chr. 14 - 123987636 - 124015684 [+] on Build GRCm38 |
