siRNA Details
Gene Symbol: 1700012M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005927.1 | - | 1 | 413 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706066.1 | - | 1 | 413 | Chr. 11 - 31870099 - 31870745 [-] on Build GRCm38 |
