siRNA Details
Gene Symbol: 1700012O15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005929.1 | - | 378 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700012O15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005929.1 | - | 378 |