siRNA Details
Gene Symbol: Gm41810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044380.1 | - | 655 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB278285.2 | - | 4 | 634 | Chr. 19 - 11818268 - 11831258 [+] on Build GRCm38 |
