siRNA Details
Gene Symbol: 6720469O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020153.1 | - | 1 | 279 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719411.1 | - | 1 | 279 | Chr. 7 - 37365441 - 37366097 [-] on Build GRCm38 |
