siRNA Details
Gene Symbol: 4930590G02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016378.1 | - | 2 | 196 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716025.1 | - | 2 | 196 | Chr. 6 - 53978701 - 53981793 [+] on Build GRCm38 |
