siRNA Details
Gene Symbol: Gm36455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041461.1 | - | 3 | 891 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB228320.2 | - | 2 | 589 | Chr. 16 - 21833850 - 21835292 [-] on Build GRCm38 |
